Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate (Powder/USP), Fisher Chemical


Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLS386212

 View more versions of this product

Catalog No. S386-212

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 282.22
Melting Point 70°C
Physical Form Powder/Solid
Quantity 2.5kg
Assay 99.0 to 102.0% (USP)
Purity Grade USP
Color White
pH 5.5 to 8.5
Molecular Formula C4H4KNaO6 · 4 H2O
Decomposition Information >100°C
Identification Pass Test (USP)
Synonym Seignette Salt
ChemAlert Storage Symbol Gray
Heavy Metals (as Pb) 0.001% max. (USP)
MDL Number MFCD00150989
Residual Solvents Meets Requirements (USP)
Water 21.0 to 27.0% (USP)
Packaging Poly Bottle
Ammonia 0.002% max. (USP)
Alkalinity Pass Test (USP)
Health Hazard 2 CAUTION!
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0
CAS 6381-59-5
Chemical Name or Material Sodium Potassium Tartrate

Specifications

Chemical Name or Material Sodium Potassium Tartrate
CAS 6381-59-5
Purity Grade USP
Physical Form Powder/Solid
Assay 99.0 to 102.0% (USP)
Molecular Formula C4H4KNaO6 · 4 H2O
Formula Weight 282.22
MDL Number MFCD00150989
Synonym Seignette Salt
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!

SDS & Safety Documents