Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate (Powder/USP), Fisher Chemical


Quantity 12kg
Packaging Poly Pail

Manufacturer: fisher scientific company  FLS38612

 View more versions of this product

Catalog No. S386-12

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 282.22
Melting Point 70°C
Quantity 12kg
Assay 99.0 to 102.0% (USP)
Purity Grade USP
Color White
Molecular Formula C4H4KNaO6 · 4 H2O
pH 5.5 to 8.5
Decomposition Information >100°C
Residual Solvents Meets Requirements (USP)
Alkalinity Pass Test (USP)
Synonym Seignette Salt
Identification Pass Test (USP)
Health Hazard 2 CAUTION!
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0
MDL Number MFCD00150989
Heavy Metals (as Pb) 0.001% max. (USP)
Water 21.0 to 27.0% (USP)
Ammonia 0.002% max. (USP)
Packaging Poly Pail
ChemAlert Storage Symbol Gray
Chemical Name or Material Sodium Potassium Tartrate
CAS 6381-59-5

Specifications

Chemical Name or Material Sodium Potassium Tartrate
CAS 6381-59-5
Purity Grade USP
Physical Form Powder/Solid
Assay 99.0 to 102.0% (USP)
Molecular Formula C4H4KNaO6 · 4 H2O
Formula Weight 282.22
MDL Number MFCD00150989
Synonym Seignette Salt
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!

SDS & Safety Documents