missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, 50% (w/v), LabChem™

Manufacturer:  LabChem LC223107

 View more versions of this product

Catalog No. LC223107



Specifications

Specifications

Sodium Potassium Tartrate
6381-59-5,7732-18-5
C4H12KNaO10
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
282.23
50% (w/v)
Passes Test
Soluble in water
Refrigerate
Colorless
For Nitrogen (Ammonia), Nitrogen (Organic)
50,50
NaKC4H4O6·4H2O
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
Liquid
Certified
Poly Bottle
Passes Test
Carbon monoxide; Carbon dioxide
125mL

Chemical Identifiers

6381-59-5
282.218
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
C4H12KNaO10
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Documents
Promotions

Promotions