missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, 10% (w/v), LabChem™

$301.74 - $301.74

Chemical Identifiers

CAS 7732-18-5,6381-59-5,7664-93-9,9002-92-0,1310-73-2
CAS Max % 89.7,10,0.2,0.05,0.05
Linear Formula NaKC4H4O6·4H2O
Formula Weight 282.23
Chemical Name or Material Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic)
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC248002 Lab Chem
LC248002
1L Each for $301.74
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6381-59-5,7664-93-9,9002-92-0,1310-73-2
NaKC4H4O6·4H2O
Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic)
89.7,10,0.2,0.05,0.05
282.23
Certified Grade

Specifications

7732-18-5,6381-59-5,7664-93-9,9002-92-0,1310-73-2
Liquid
282.23
Poly Bottle
Refrigerate
Clear, colorless
Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic)
Certified Grade
89.7,10,0.2,0.05,0.05
NaKC4H4O6·4H2O
Passes Test
Soluble in water
Carbon monoxide; Carbon dioxide
1L
10% (w/v)

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.