Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate, For Nitrogen (Ammonia), Certified, 30% (w/v), LabChem™

$92.51 - $92.51

Chemical Identifiers

Chemical Name or Material Sodium Potassium Tartrate
CAS 7732-18-5
CAS Max % 70
CAS 6381-59-5
CAS Max % 30
Name Note For Nitrogen (Ammonia)
Purity Grade Certified Grade
Linear Formula NaKC4H4O6·4H2O
Formula Weight 282.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC248301

 
lab chem
LC248301
500mL Each for $92.51
Description & Specifications

Specifications

Formula Weight 282.23
Purity Grade Certified Grade
CAS 6381-59-5
Suitability Passes Test
Assay 30% (w/v)
Storage Information Refrigerate
Decomposition Information Carbon monoxide; Carbon dioxide
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS 7732-18-5
Packaging Poly Bottle
CAS Max % 30
Solubility Soluble in water
Linear Formula NaKC4H4O6·4H2O
Name Note For Nitrogen (Ammonia)
Physical Form Liquid
Color Clear, colorless
CAS Max % 70
Quantity 500mL
Identification Passes Test
Chemical Name or Material Sodium Potassium Tartrate
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.