missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium, plasma standard solution, Na 10,000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 14400AD

 View more versions of this product

Catalog No. AA14400AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium, plasma standard solution
Na2CO3 in 5% HNO3
Nitric acid: 5%; Sodium carbonate: 2.3%; Water: 92.7%
23 hPa (17mm Hg) at 20°C
50mL
Clear liquid
MFCD00085307
Liquid
Undesignated

Chemical Identifiers

Na2CO3 in 5% HNO3
MFCD00085307
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

Documents