Learn More
Sodium phosphonoformate hexahydrate, 98+%, Thermo Scientific Chemicals
$182.42 - $1748.66
Chemical Identifiers
CAS | 34156-56-4 |
---|---|
Molecular Formula | CH12Na3O11P |
Molecular Weight (g/mol) | 300.04 |
MDL Number | MFCD00150176 |
InChI Key | ILRVASBWNRYBFD-UHFFFAOYSA-K |
Synonym | foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate |
PubChem CID | 169569 |
ChEBI | CHEBI:60269 |
SMILES | O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2478403
|
Thermo Scientific Chemicals
B2478403 |
1 g |
Each for $182.42
|
|
|||||
AAB2478406
|
Thermo Scientific Chemicals
B2478406 |
5 g |
Each for $523.30
|
|
|||||
AAB2478414
|
Thermo Scientific Chemicals
B2478414 |
25 g |
Each for $1,748.66
|
|
|||||
Description
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
34156-56-4 | |
300.04 | |
ILRVASBWNRYBFD-UHFFFAOYSA-K | |
169569 | |
O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O |
CH12Na3O11P | |
MFCD00150176 | |
foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate | |
CHEBI:60269 |
Specifications
34156-56-4 | |
MFCD00150176 | |
foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate | |
ILRVASBWNRYBFD-UHFFFAOYSA-K | |
trisodium;phosphonatoformate;hexahydrate | |
169569 | |
300.08 (191.98 Anhydrous) | |
Sodium phosphonoformate hexahydrate |
CH12Na3O11P | |
1 g | |
Soluble in water. | |
O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O | |
300.04 | |
CHEBI:60269 | |
≥98% |
Safety and Handling
Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
RTECSNumber : SY8200000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only