missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium phosphonoformate hexahydrate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2478414

 View more versions of this product

Catalog No. AAB2478414



Description

Description

Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

Sodium phosphonoformate hexahydrate
CH12Na3O11P
25 g
Soluble in water.
O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O
300.04
CHEBI:60269
≥98%
34156-56-4
MFCD00150176
foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate
ILRVASBWNRYBFD-UHFFFAOYSA-K
trisodium hexahydrate phosphonatoformate
169569
300.08 (191.98 Anhydrous)

Chemical Identifiers

34156-56-4
300.04
ILRVASBWNRYBFD-UHFFFAOYSA-K
169569
O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O
CH12Na3O11P
MFCD00150176
foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate
CHEBI:60269
Safety and Handling

Safety and Handling

Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation

Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)

RTECSNumber : SY8200000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only