Please login to your online account to display your discounted pricing

Sodium phosphite dibasic pentahydrate, 98%, Acros Organics


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428500010

 View more versions of this product

Catalog No. AC428500010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium phosphite dibasic pentahydrate
CAS 13517-23-2
Assay Percent Range 98%
Molecular Formula HNa2O35H2O
Formula Weight 216.02
Synonym di-Sodium hydrogen phosphite pentahydrate
Packaging Glass bottle
Quantity 1kg

Specifications

Chemical Name or Material Sodium phosphite dibasic pentahydrate
Assay Percent Range 98%
Molecular Formula HNa2O35H2O
Formula Weight 216.02
Synonym di-Sodium hydrogen phosphite pentahydrate