Learn More
Sodium phosphate, tribasic, pure, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389810010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium phosphate, tribasic | |
7601-54-9 | |
Crystalline Powder | |
Na3O4P | |
MFCD00003510 | |
trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat | |
RYFMWSXOAZQYPI-UHFFFAOYSA-K | |
trisodium;phosphate | |
24243 | |
163.94 | |
Pure |
Anhydrous, Pure | |
White | |
1 kg | |
Na3PO4 | |
15, 8792 | |
Solubility in water: 88g/L (20°C) | |
[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] | |
163.94 | |
CHEBI:37583 | |
≥96.0% | |
Plastic Bottle |
Chemical Identifiers
7601-54-9 | |
163.94 | |
RYFMWSXOAZQYPI-UHFFFAOYSA-K | |
24243 | |
trisodium;phosphate |
Na3O4P | |
MFCD00003510 | |
trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat | |
CHEBI:37583 | |
[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
EINECSNumber : 231-509-8
RUO – Research Use Only