Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic Dodecahydrate, MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  0215206280

 View more versions of this product

Catalog No. ICN15206280

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
CAS 10101-89-0
Molecular Formula Na3PO4 · 12H2O
Formula Weight 380.1g/mol
Synonym (Trisodium phosphate)
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
CAS 10101-89-0
Molecular Formula Na3PO4 · 12H2O
Formula Weight 380.1g/mol
Synonym (Trisodium phosphate)
Accessories

Room Temperature