Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic Dodecahydrate, MP Biomedicals

$45.26 - $45.26

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
CAS 10101-89-0
Molecular Formula Na3PO4 · 12H2O
Formula Weight 380.1
Synonym (Trisodium phosphate)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15206280

 
mp biomedicals
15206280
100g Each for $45.26
Description & Specifications

Specifications

Formula Weight 380.1
Quantity 100g
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
Molecular Formula Na3PO4 · 12H2O
CAS 10101-89-0
Synonym (Trisodium phosphate)
Recommended Storage Room Temperature