Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic Dodecahydrate (Granular/Technical), Fisher Chemical


Quantity 50kg
Packaging Fiber Drum

Manufacturer: fisher scientific company  S37650

 View more versions of this product

Catalog No. S376-50

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 380.12
Melting Point 75°C
Quantity 50kg
Physical Form Solid
Assay 99 to 122%
Molecular Formula Na3O4P · 12 H2O
Viscosity [NULL]
Color White
pH 11.8 to 12
Density 1.620g/cm³
Packaging Fiber Drum
Synonym Trisodium Phosphate
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
Identification Pass Test
Assay Percent Range 99 to 122 %
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Causes severe burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:1
Instability:0
MDL Number MFCD00149198
ChemAlert Storage Symbol Gray
CAS 10101-89-0

Specifications

Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
CAS 10101-89-0
Purity Grade Technical Grade
Physical Form Solid
Assay 99 to 122%
Assay Percent Range 99 to 122 %
Molecular Formula Na3O4P · 12 H2O
Formula Weight 380.12
MDL Number MFCD00149198
Synonym Trisodium Phosphate
Accessories
 

Emergency Overview
Causes severe burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:1
Instability:0

DANGER!

SDS & Safety Documents