missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic Dodecahydrate (Granular/Technical), Fisher Chemical

Manufacturer:  Fisher Scientific Company S3763

 View more versions of this product

Catalog No. S376-3



 

Specifications Description & Specifications

Specifications

Sodium Phosphate Tribasic Dodecahydrate
Technical Grade
99 to 122%
Na3O4P · 12 H2O
MFCD00149198
1.620g/cm³
Poly Bottle
75°C
3kg
10101-89-0
Solid
99 to 122 %
380.12
Trisodium Phosphate
Pass Test
White
11.8 to 12

Chemical Identifiers

10101-89-0
99 to 122 %
380.12
Trisodium Phosphate
99 to 122%
Solid
Na3O4P · 12 H2O
MFCD00149198
Sodium Phosphate Tribasic Dodecahydrate
Technical Grade

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Causes severe burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:1
Instability:0