Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic Dodecahydrate (Granular/Technical), Fisher Chemical

$422.86 - $6,004.57

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
CAS 10101-89-0
Purity Grade Technical Grade
Physical Form Solid
Assay 99 to 122%
Assay Percent Range 99 to 122 %
Molecular Formula Na3O4P · 12 H2O
Formula Weight 380.12
MDL Number MFCD00149198
Synonym Trisodium Phosphate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S376-3

  SDS
fisher scientific company
S3763
Poly Bottle 3kg Each for $422.86
Case of 4 Each for $1,691.44

S376-50

  SDS
fisher scientific company
S37650
Fiber Drum 50kg Each for $6,004.57
Description & Specifications

Specifications

Melting Point 75°C
Purity Grade Technical Grade
Formula Weight 380.12
Quantity 3kg
Assay 99 to 122%
Physical Form Solid
Color White
pH 11.8 to 12
Molecular Formula Na3O4P · 12 H2O
Density 1.620g/cm³
Identification Pass Test
Assay Percent Range 99 to 122 %
Health Hazard 2 DANGER!
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
CAS 10101-89-0
MDL Number MFCD00149198
Packaging Poly Bottle
Synonym Trisodium Phosphate
Health Hazard 3 Emergency Overview
Causes severe burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:1
Instability:0
ChemAlert Storage Symbol Gray

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Causes severe burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:1
Instability:0

DANGER!