Learn More
Sodium Phosphate Tribasic Dodecahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific S3773
Specifications
| Sodium Phosphate Tribasic Dodecahydrate | |
| 10101-89-0 | |
| 11.8 to 12 | |
| 3 kg | |
| H24Na3O16P | |
| trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
| O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] | |
| 380.119 | |
| 380.12 | |
| Certified ACS | |
| Poly Bottle |
| 75°C | |
| White | |
| Solid | |
| 98 to 102 % | |
| MFCD00149198 | |
| ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
| trisodium;phosphate;dodecahydrate | |
| 61473 | |
| 98 to 102% | |
| Pass Test | |
| 1.620g/cm³ |
Chemical Identifiers
| 10101-89-0 | |
| 380.119 | |
| ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
| 61473 | |
| O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
| H24Na3O16P | |
| MFCD00149198 | |
| trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
| trisodium;phosphate;dodecahydrate |
Safety and Handling
DANGER!
Emergency Overview
Causes severe burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .
NFPA
Health:3
Flammability:1
Instability:0