Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic, Dodecahydrate ACS, EMD Millipore


Quantity 12kg
Packaging Poly Pail

Manufacturer: emd millipore corporation  SX07257

 View more versions of this product

Catalog No. MSX07257

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 380.12
Quantity 12kg
Chemical Name or Material Sodium Phosphate
Name Note Tribasic, Dodecahydrate
Packaging Poly Pail
Linear Formula Na3PO4·12H2O
CAS 10101-89-0

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Tribasic, Dodecahydrate
CAS 10101-89-0
Purity Grade ACS Grade
Linear Formula Na3PO4·12H2O
Formula Weight 380.12