Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic, Dodecahydrate ACS, EMD Millipore


Quantity 500g
Packaging Poly Bottle

Manufacturer: emd millipore corporation  SX07251

 View more versions of this product

Catalog No. MSX07251

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 380.12
Purity Grade ACS Grade
Quantity 500g
Name Note Tribasic, Dodecahydrate
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate
Linear Formula Na3PO4·12H2O
CAS 10101-89-0

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Tribasic, Dodecahydrate
CAS 10101-89-0
Purity Grade ACS Grade
Linear Formula Na3PO4·12H2O
Formula Weight 380.12