Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic, Dodecahydrate ACS, EMD Millipore


Quantity 50kg
Packaging Fiber Drum w/bag

Manufacturer: emd millipore corporation  SX072520

 View more versions of this product

Catalog No. MSX072520

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 380.12
Purity Grade ACS Grade
Quantity 50kg
CAS 10101-89-0
Name Note Tribasic, Dodecahydrate
Chemical Name or Material Sodium Phosphate
Packaging Fiber Drum w/bag
Linear Formula Na3PO4·12H2O

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Tribasic, Dodecahydrate
CAS 10101-89-0
Purity Grade ACS Grade
Linear Formula Na3PO4·12H2O
Formula Weight 380.12