missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic, Dodecahydrate ACS, EMD Millipore

$138.17 - $2,892.09

Chemical Identifiers

CAS 10101-89-0
Linear Formula Na3PO4·12H2O
Formula Weight 380.12
Chemical Name or Material Sodium Phosphate, Tribasic, Dodecahydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MSX07251 Emd Millipore Corporation
SX07251
Poly Bottle 500g Each for $138.17
MSX07257 Emd Millipore Corporation
SX07257
Poly Pail 12kg Each for $1,017.46
MSX072520 Emd Millipore Corporation
SX072520
Fiber Drum w/bag 50kg Each for $2,892.09
Specifications Description & Specifications

Chemical Identifiers

10101-89-0
380.12
ACS Grade
Na3PO4·12H2O
Sodium Phosphate, Tribasic, Dodecahydrate

Specifications

380.12
Poly Bottle
10101-89-0
ACS Grade
500g
Na3PO4·12H2O
Sodium Phosphate, Tribasic, Dodecahydrate