Learn More
Sodium phosphate, tribasic dodecahydrate, extra pure, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC450660250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium phosphate, tribasic dodecahydrate | |
75.0°C | |
Na3O4P·12H2O | |
15,8792 | |
Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] | |
380.13 | |
380.13 | |
0.05% max. (in water) | |
Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol | |
1.6200g/mL |
10101-89-0 | |
25kg | |
Na3PO4·12H2O | |
ccris 7322, phosphoric acid, trisodium salt, dodeahydrate, phosphoric acid, trisodium salt, dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, trisodium phosphate dodecahydrate, trisodium phosphate tert dodecahydrate, unii-b70850qphr | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
trisodium;phosphate;dodecahydrate | |
61473 | |
Extra Pure | |
Fibre drum | |
1.62 |
Chemical Identifiers
10101-89-0 | |
380.13 | |
ccris 7322, phosphoric acid, trisodium salt, dodeahydrate, phosphoric acid, trisodium salt, dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, trisodium phosphate dodecahydrate, trisodium phosphate tert dodecahydrate, unii-b70850qphr | |
trisodium;phosphate;dodecahydrate |
Na3O4P·12H2O | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
61473 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
RUO – Research Use Only