missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Sodium phosphate, tribasic dodecahydrate, 98%, for biochemistry
Chemical Identifiers
CAS | 10101-89-0 |
---|---|
Molecular Formula | Na3O4P·12H2O |
Molecular Weight (g/mol) | 380.13 |
InChI Key | ASTWEMOBIXQPPV-UHFFFAOYSA-K |
Synonym | trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate |
PubChem CID | 61473 |
IUPAC Name | trisodium;phosphate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC447965000
|
Thermo Scientific™
447965000 |
500g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC447960025
|
Thermo Scientific™
447960025 |
2.5kg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Biological bufferChemical Identifiers
10101-89-0 | |
380.13 | |
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
trisodium;phosphate;dodecahydrate |
Na3O4P·12H2O | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
61473 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Specifications
10101-89-0 | |
98% | |
Na3PO4·12H2O | |
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
trisodium;phosphate;dodecahydrate | |
61473 | |
98% | |
Plastic bottle | |
1.6200g/mL |
500g | |
Na3O4P·12H2O | |
15,8792 | |
Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] | |
380.13 | |
380.13 | |
Biochemistry | |
1.62 | |
Sodium phosphate, tribasic dodecahydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
RUO â Research Use Only