Sodium Phosphate Tribasic Dodecahydrate, 98.0-102.0%, (ACS Reagent Grade), MP Biomedicals
Manufacturer:  Mp Biomedicals 15257890
 View more versions of this product
Catalog No. ICN15257890
Specifications Description & Specifications
Specifications
380.1
500g
Na3PO4 · 12H2O
(Trisodium phosphate)
ACS Reagent Grade
10101-89-0
98.0-102.0%
Sodium Phosphate Tribasic Dodecahydrate
Chemical Identifiers
10101-89-0
Na3PO4 · 12H2O
(Trisodium phosphate)
ACS Reagent Grade
98.0-102.0%
380.1
Sodium Phosphate Tribasic Dodecahydrate
Safety and Handling
Recommended Storage : Room Temperature