Please login to your online account to display your discounted pricing

Sodium Phosphate Tribasic Dodecahydrate, 98.0-102.0%, (ACS Reagent Grade), MP Biomedicals

$114.79 - $114.79

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
CAS 10101-89-0
Purity Grade ACS Reagent Grade
Assay Percent Range 98.0-102.0%
Molecular Formula Na3PO4 · 12H2O
Formula Weight 380.1
Synonym (Trisodium phosphate)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15257890

 
mp biomedicals
15257890
500g Each for $114.79
Description & Specifications

Specifications

Formula Weight 380.1
Purity Grade ACS Reagent Grade
Quantity 500g
CAS 10101-89-0
Molecular Formula Na3PO4 · 12H2O
Recommended Storage Room Temperature
Assay Percent Range 98.0-102.0%
Synonym (Trisodium phosphate)
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
Safety and Handling

Room Temperature