missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium phosphate, tribasic, anhydrous, tech, Alfa Aesar™

Manufacturer:  Alfa Aesar 13438A1

 View more versions of this product

Catalog No. AA13438A1



Specifications

Specifications

Sodium phosphate, tribasic, anhydrous
Na3O4P
White powder
RYFMWSXOAZQYPI-UHFFFAOYSA-K
trisodium;phosphate
24243
163.94
Technical
Odorless
1kg
7601-54-9
MFCD00003510
trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
163.939
CHEBI:37583
Powder
1.62g/cm3
White

Chemical Identifiers

7601-54-9
163.939
RYFMWSXOAZQYPI-UHFFFAOYSA-K
24243
trisodium;phosphate
Na3O4P
MFCD00003510
trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
CHEBI:37583
[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-509-8

SDS Documents