Please login to your online account to display your discounted pricing

Sodium Phosphate, Tribasic, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$857.95 - $3,178.98

Chemical Identifiers

Chemical Name or Material Sodium Phosphate
CAS 10101-89-0
Name Note Tribasic, 12-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na3PO4·12H2O
Formula Weight 380.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004196

 
avantor performance
383601
Poly Bottle 500g Case of 4 Each for $857.95

02004197

 
avantor performance
383605
Poly Bottle 2.5kg Case of 4 Each for $3,178.98
Description & Specifications

Specifications

Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 380.12
CAS 10101-89-0
Chemical Name or Material Sodium Phosphate
Molecular Formula Na3PO4·12H2O
Name Note Tribasic, 12-Hydrate
Quantity 500g
Specific Gravity 1L = 1.62kg
Packaging Poly Bottle