Please login to your online account to display your discounted pricing

Sodium Phosphate, Tribasic, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  383605

 View more versions of this product

Catalog No. 02004197

  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 380.12
Chemical Name or Material Sodium Phosphate
Molecular Formula Na3PO4·12H2O
Name Note Tribasic, 12-Hydrate
Packaging Poly Bottle
Specific Gravity 1L = 1.62kg
CAS 10101-89-0
Quantity 2.5kg

Specifications

Chemical Name or Material Sodium Phosphate
CAS 10101-89-0
Name Note Tribasic, 12-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na3PO4·12H2O
Formula Weight 380.12