Please login to your online account to display your discounted pricing

Sodium Phosphate Solution, Saturated, for Chlorine Dioxide, LabChem™

$103.29 - $103.29

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Solution
CAS 7732-18-5
CAS Max % 88
CAS 7558-79-4
CAS Max % 12
Name Note Saturated, for Chlorine Dioxide
Linear Formula Na2HPO4
Formula Weight 141.96
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC247802

 
lab chem
LC247802
1L Each for $103.29
Description & Specifications

Specifications

Formula Weight 141.96
CAS 7558-79-4
Health Hazard 2 GHS H Statement
Causes eye irritation.
Chemical Name or Material Sodium Phosphate Solution
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
CAS Max % 88
Identification Passes Test
Quantity 1L
Name Note Saturated, for Chlorine Dioxide
CAS 7732-18-5
CAS Max % 12
Color Clear colorless
Health Hazard 1 Warning
Packaging Poly Bottle
Solubility Soluble in water
Physical Form Liquid
Linear Formula Na2HPO4
Storage Information Room Temperature
Decomposition Information Phosphorus oxides
Safety and Handling

Warning

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes eye irritation.