Please login to your online account to display your discounted pricing

Sodium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

$735.04 - $1,065.20

Chemical Identifiers

Chemical Name or Material Sodium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003952

 
avantor performance
D02104
Narrow Mouth Polyethylene Bottle 1.9L Each for $1,065.20

02003953

 
avantor performance
D02100
Poly Bottle 230.8mL Each for $879.74

02003954

 
avantor performance
D02102
Narrow Mouth Polyethylene Bottle 961.5mL Each for $735.04
Description & Specifications

Specifications

Quantity 1.9L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Sodium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate
Packaging Narrow Mouth Polyethylene Bottle