Please login to your online account to display your discounted pricing

Sodium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™


Quantity 230.8mL
Packaging Poly Bottle

Manufacturer: avantor performance  D02100

 View more versions of this product

Catalog No. 02003953

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Quantity 230.8mL

Specifications

Chemical Name or Material Sodium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate