Please login to your online account to display your discounted pricing

Sodium Phosphate, Monobasic, Monohydrate, Crystal, Ultrapure Bioreagent, J.T.Baker™


High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  401105

 View more versions of this product

Catalog No. 02004218

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 137.99
Physical Form Crystals
CAS 10049-21-5
Chemical Name or Material Sodium Phosphate
Purity Grade Ultrapure Bioreagent
Molecular Formula NaH2PO4·H2O
Name Note Monobasic, Monohydrate
Packaging Poly Bottle
Quantity 2.5kg
Specific Gravity 1L = 2.04kg

Specifications

Chemical Name or Material Sodium Phosphate
CAS 10049-21-5
Name Note Monobasic, Monohydrate
Purity Grade Ultrapure Bioreagent
Molecular Formula NaH2PO4·H2O
Formula Weight 137.99

    For Liquid Chromatography and Molecular Biology applications