missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals), Fisher BioReagents

$53.33 - $79.70

Chemical Identifiers

CAS 10049-21-5
Physical Form Solid
Molecular Formula H2NaO4P · H2O
Formula Weight 137.99
Synonym Sodium Dihydrogen Phosphate
Chemical Name or Material Sodium Phosphate Monobasic Monohydrate
Assay 98.0 to 102.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BP330-500 SDS Fisher Scientific Company
BP330-500
Amber Glass 500g Each for $53.33
Case of 6 Each for $319.98
BP330-1 SDS Fisher Scientific Company
BP3301
Amber Glass 1kg Each for $79.70
Case of 6 Each for $478.20
Specifications

Description

Sodium Phosphate is commonly used in tissue culture and molecular biology applications.
Description & Specifications

Chemical Identifiers

10049-21-5
H2NaO4P · H2O
Sodium Dihydrogen Phosphate
98.0 to 102.0%
Solid
137.99
Sodium Phosphate Monobasic Monohydrate

Specifications

100°C
500g
H2NaO4P · H2O
White
0.003% max.
Sodium Dihydrogen Phosphate
Pass Test
Amber Glass
0.001% max.
Sodium Phosphate Monobasic Monohydrate
137.99
Solid
4.1 to 4.5
0.01% max.
0.01% max.
10049-21-5
5ppm max.
0.005% max.
0.001% max.
98.0 to 102.0%

Safety and Handling

Recommended Storage : RT