missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phosphate, monobasic monohydrate, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207805000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
Sodium phosphate, monobasic monohydrate | |
p.a. | |
White | |
Crystalline Powder and/or Crystals | |
99+% | |
NaH2PO4·H2O | |
01,395 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium phosphoric acid hydrate | |
516949 | |
≥99% | |
Plastic bottle |
100.0°C | |
10049-21-5 | |
4.1 to 4.5 (5% soln. at 20°C) | |
500 g | |
H5NaO5P | |
MFCD00149208 | |
15, 8790 | |
Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.[Na+].OP(O)(O)=O | |
139.00 | |
137.99 | |
Analytical |
Chemical Identifiers
10049-21-5 | |
139.00 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only