Please login to your online account to display your discounted pricing

Sodium phosphate, monobasic monohydrate, 98+%, ACS reagent, ACROS Organics™


Quantity 10kg
Packaging Plastic bucket

Manufacturer: thermo fisher scientific  AC424390100

 View more versions of this product

Catalog No. AC424390100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Color White semi-transparentor
Physical Form Crystals or Crystalline Powder
Formula Weight 137.99
Quantity 10kg
Assay 98.0 to 102.0%
pH 4.1 to 4.5 (5% soln. at 25°C)
Potassium (K) 100ppm max.
Insoluble Matter 0.01% max.
Linear Formula NaH2PO4·H2O
Iron (Fe) 10ppm max.
Chemical Name or Material Sodium phosphate, monobasic monohydrate
CAS 10049-21-5
Assay Percent Range 98+%
Calcium (Ca) 50ppm max.
Molecular Formula H2NaO4P·H2O
Heavy Metals (as Pb) 10ppm max.
Packaging Plastic bucket
Sulfate 30ppm max.
Synonym Dihydrogen sodium phosphate monohydrate
Chlorine (Cl) 5ppm max.

Specifications

Chemical Name or Material Sodium phosphate, monobasic monohydrate
CAS 10049-21-5
Purity Grade ACS Reagent Grade
Assay 98.0 to 102.0%
Assay Percent Range 98+%
Molecular Formula H2NaO4P·H2O
Linear Formula NaH2PO4·H2O
Formula Weight 137.99
Synonym Dihydrogen sodium phosphate monohydrate