missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phosphate, monobasic dihydrate, 99+%, for analysis, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 271750100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium phosphate, monobasic dihydrate | |
10kg | |
H2NaO4P·2H2O | |
MFCD00149209 | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate;dihydrate | |
23673460 | |
99+% | |
24% max. |
13472-35-0 | |
99+% | |
NaH2PO4·2H2O | |
15, 8790 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
O.O.OP(=O)(O)[O-].[Na+] | |
156.01 | |
156.01 | |
Analysis | |
Plastic drum |
RUO â Research Use Only