missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate, Monobasic, Anhydrous, Reagent Grade, LabChem™

$103.34 - $103.34

Chemical Identifiers

CAS 7558-80-7
CAS Max % 100
Linear Formula NaH2PO4
Formula Weight 119.98
Chemical Name or Material Sodium Phosphate, Monobasic, Anhydrous
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC247751 Lab Chem
LC247751
500g Each for $103.34
Specifications Description & Specifications

Chemical Identifiers

7558-80-7
NaH2PO4
Sodium Phosphate, Monobasic, Anhydrous
100
119.98
Reagent Grade

Specifications

7558-80-7
Crystals or Granules
119.98
Passes Test
Soluble in water
Room Temperature
White
500g
Reagent Grade
100
NaH2PO4
2.04g/cm3
Poly Bottle
2.04g/cm3
Phosphorus oxides
203.9°C
Sodium Phosphate, Monobasic, Anhydrous

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

EINECSNumber : 231-449-2