missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™
For tissue culture and molecular biology applications
Supplier: Fisher BioReagents FLBP3291
Description
For tissue culture and molecular biology applications
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.Specifications
203.9°C | |
100°C | |
White | |
Solid | |
≥99 % | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
≥99% | |
Poly Bottle |
Sodium Phosphate Monobasic Anhydrous | |
7558-80-7 | |
4.0 - 6.0 | |
1 kg | |
H2NaO4P | |
15, 8790 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium dihydrogen phosphate | |
23672064 | |
119.98 | |
Pass Test | |
2.04g/cm³ |
Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
23672064 | |
[Na+].OP(O)([O-])=O |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
CHEBI:37585 |
Safety and Handling
Recommended Storage : RT