missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phosphate, monobasic, 99%, for analysis, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389870010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
Sodium phosphate, monobasic | |
7558-80-7 | |
Crystalline Powder | |
99% | |
NaH2PO4 | |
15,879 | |
Solubility in water: 850g/L (20°C). | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
99% | |
Plastic bottle |
White | |
4.1 to 4.7 (5% soln.) | |
1 kg | |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate | |
23672064 | |
119.98 | |
Analytical |
Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
23672064 | |
[Na+].OP(O)([O-])=O |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
CHEBI:37585 |
Safety and Handling
EINECSNumber : 231-449-2
RUO – Research Use Only