Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic, (White, crystalline powder/ACS Reagent Grade), MP Biomedicals

$126.55 - $126.55

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Dibasic
CAS 7558-79-4
Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder
Molecular Formula Na2HPO4
Formula Weight 141.96
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19144001

 
mp biomedicals
19144001
1kg Each for $126.55
Description & Specifications

Specifications

Formula Weight 141.96
Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder
Color White
Quantity 1kg
CAS 7558-79-4
Chemical Name or Material Sodium Phosphate Dibasic
Molecular Formula Na2HPO4