Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic, MP Biomedicals


Quantity 1kg

Manufacturer: mp biomedicals  0219484991

 View more versions of this product

Catalog No. ICN19484991

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1kg
Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
CAS 7558-79-4
Molecular Formula Na2HPO4
Formula Weight 141.96g/mol

Specifications

Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
CAS 7558-79-4
Molecular Formula Na2HPO4
Formula Weight 141.96g/mol