missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic, MP Biomedicals

$93.14 - $152.81

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19484983
View Documents Promotion Details
MP Biomedicals Inc
0219484983
250 g
Each for $93.14
 
ICN19484990
View Documents Promotion Details
MP Biomedicals Inc
0219484990
500 g
Each for $152.81
 
Description

Description

Specifications

Chemical Identifiers

7558-79-4
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
disodium hydrogen phosphate
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O

Specifications

7558-79-4
HNa2O4P
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
[Na+].[Na+].OP([O-])([O-])=O
141.96
CHEBI:34683
Sodium Phosphate Dibasic, Anhydrous
250 g
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
24203
141.96
SDS
missing translation for 'documents'

missing translation for 'documents'