Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic Heptahydrate (Granular/USP), Fisher Chemical

$508.29 - $508.29

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
CAS 7782-85-6
Purity Grade USP
Physical Form Solid
Assay 98.0 to 100.5% (USP)
Molecular Formula HNa2O4P · 7 H2O
Formula Weight 268.07
MDL Number MFCD00149180
Synonym Disodium Hydrogen Phosphate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S471-3

  SDS
fisher scientific company
S4713
Poly Bottle 3kg (6.6 lb.) Each for $508.29
Case of 4 Each for $2,033.16
Description & Specifications

Specifications

Melting Point 48°C
Formula Weight 268.07
Quantity 3kg (6.6 lb.)
Assay 98.0 to 100.5% (USP)
Molecular Formula HNa2O4P · 7 H2O
Physical Form Solid
Purity Grade USP
pH 9.0 to 9.3
Color White
Sulfate 0.2% max. (USP)
Chloride 0.06% max. (USP)
Insoluble Matter 0.4% max. (USP)
Heavy Metals (as Pb) 0.002% max. (USP)
Residual Solvents Meets Requirements (USP)
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
ChemAlert Storage Symbol Gray
MDL Number MFCD00149180
Loss on Drying 43.0 to 50.0% (USP)
Arsenic (As) 16ppm max. (USP)
Identification Pass Test (USP)
Synonym Disodium Hydrogen Phosphate
CAS 7782-85-6

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling