Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals), Fisher BioReagents

$98.75 - $162.83

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
CAS 7782-85-6
Physical Form Solid
Assay 98.0 to 102.0%
Assay Percent Range 98.0 to 102.0 %
Assay Percent Notes 99.0-102.0%
Molecular Formula HNa2O4P · 7 H2O
Formula Weight 268.07
Synonym Disodium Hydrogen Phosphate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP331-1

  SDS
fisher scientific company
BP3311
Poly Bottle 1kg Each for $162.83
Case of 6 Each for $976.98

BP331-500

  SDS
fisher scientific company
FLBP331500
Glass Bottle 500g Each for $98.75
Case of 6 Each for $592.50
Description & Specifications

Specifications

Formula Weight 268.07
Melting Point 48°C
Quantity 1kg
Physical Form Solid
Assay 98.0 to 102.0%
Molecular Formula HNa2O4P · 7 H2O
Color White
pH 9.0 to 9.3
Merck Index 15, 8789
Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
Iron (Fe) 0.001% max.
Assay Percent Range 98.0 to 102.0 %
Sulfate 0.005% max.
CAS 7782-85-6
Synonym Disodium Hydrogen Phosphate
Recommended Storage RT
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Insoluble Matter 0.005% max.
Packaging Poly Bottle
Assay Percent Notes 99.0-102.0%
ChemAlert Storage Symbol Gray
Chloride 0.001% max.

This buffer component is commonly used in tissue culture and molecular biology applications.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

RT