missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phosphate, dibasic heptahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 206515000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
Sodium phosphate, dibasic heptahydrate | |
p.a. | |
White | |
Crystals | |
99+% | |
Na2HPO4·7H2O | |
15,8789 | |
Solubility in water: 154g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O | |
270.08 | |
268.07 | |
Analytical | |
1.67 |
48.0°C | |
7782-85-6 | |
9.0 to 9.3 (5% soln. at 20°C) | |
500 g | |
H17Na2O11P | |
MFCD00149180 | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
disodium;hydrogen phosphate;heptahydrate | |
6096963 | |
≥99% | |
Plastic bottle |
Chemical Identifiers
7782-85-6 | |
270.08 | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
6096963 | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O |
H17Na2O11P | |
MFCD00149180 | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
disodium;hydrogen phosphate;heptahydrate |
Safety and Handling
HYGROSCOPIC
RTECSNumber : WC4600000
RUO – Research Use Only