Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic Heptahydrate, 98.0-102.0%, (White, crystalline powder/ACS Reagent Grade), MP Biomedicals

$178.65 - $178.65

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
CAS 7782-85-6
Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder
Assay Percent Range 98.0-102.0%
Molecular Formula Na2HPO4 · 7H2O
Formula Weight 268.07
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19144101

 
mp biomedicals
19144101
1kg Each for $178.65
Description & Specifications

Specifications

Physical Form Crystalline Powder
Purity Grade ACS Reagent Grade
Formula Weight 268.07
Color White
Quantity 1kg
CAS 7782-85-6
Molecular Formula Na2HPO4 · 7H2O
Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
Recommended Storage Room Temperature
Assay Percent Range 98.0-102.0%
Safety and Handling

Room Temperature