missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic dihydrate, meets analytical spec. of BP, Ph. Eur., 98.5-101% (calc. to the dried substance), Honeywell Fluka™
Meets analytical specification of BP, Ph. Eur., 98.5-101% (calc. to the dried substance)
Supplier: Honeywell-Fluka 042724X5KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Phosphate Dibasic Dihydrate | |
| 10028-24-7 | |
| H5Na2O6P | |
| MFCD00149182 | |
| KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
| disodium;hydrogen phosphate;dihydrate | |
| 6451167 | |
| 98.5 to 101% (calc. to the dried substance) |
| Meets analytical specification of Ph.Eur.BP | |
| 4 x 5 kg | |
| Na2HPO4 · 2H2O | |
| disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
| O.O.[Na+].[Na+].OP([O-])([O-])=O | |
| 177.99 | |
| 177.99 |
Chemical Identifiers
| 10028-24-7 | |
| 177.99 | |
| KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
| 6451167 |
| H5Na2O6P | |
| MFCD00149182 | |
| disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
| O.O.[Na+].[Na+].OP([O-])([O-])=O |
Safety and Handling
EINECSNumber : 231-448-7