missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phosphate dibasic dihydrate, 99+%, for HPLC, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 343810250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium phosphate dibasic dihydrate | |
For HPLC | |
10028-24-7 | |
Crystalline Powder and/or Crystals | |
99+% | |
Na2HPO4·2H2O | |
15,8789 | |
Solubility in water: 93g/L(20°C) | |
O.O.[Na+].[Na+].OP([O-])([O-])=O | |
177.99 | |
177.99 | |
HPLC |
243.0°C to 245.0°C | |
White | |
7.5 to 9.2 (5% soln. at 25°C) | |
25 g | |
H5Na2O6P | |
MFCD00149182 | |
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
disodium;hydrogen phosphate;dihydrate | |
6451167 | |
≥99% | |
Glass bottle |
Chemical Identifiers
10028-24-7 | |
177.99 | |
KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
6451167 | |
O.O.[Na+].[Na+].OP([O-])([O-])=O |
H5Na2O6P | |
MFCD00149182 | |
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
disodium;hydrogen phosphate;dihydrate |
RUO – Research Use Only