Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic, (Cell Culture Reagent), MP Biomedicals


Quantity 5kg

Manufacturer: mp biomedicals  0219473905

 View more versions of this product

Catalog No. ICN19473905

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5kg
Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
CAS 7558-79-4
Purity Grade Cell Culture Reagent
Molecular Formula Na2HPO4
Formula Weight 141.96g/mol
Synonym (Disodium hydrogen phosphate)
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
CAS 7558-79-4
Purity Grade Cell Culture Reagent
Molecular Formula Na2HPO4
Formula Weight 141.96g/mol
Synonym (Disodium hydrogen phosphate)
Accessories

Room Temperature