Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic, (Cell Culture Reagent), MP Biomedicals


Quantity 1kg

Manufacturer: mp biomedicals  19473901

 View more versions of this product

Catalog No. ICN19473901

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Cell Culture Reagent Grade
Formula Weight 141.96
Quantity 1kg
Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
Synonym (Disodium hydrogen phosphate)
Molecular Formula Na2HPO4
CAS 7558-79-4
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
CAS 7558-79-4
Purity Grade Cell Culture Reagent Grade
Molecular Formula Na2HPO4
Formula Weight 141.96
Synonym (Disodium hydrogen phosphate)
Accessories

Room Temperature