Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic, (Cell Culture Reagent), MP Biomedicals

$156.23 - $156.23

Chemical Identifiers

Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
CAS 7558-79-4
Purity Grade Cell Culture Reagent Grade
Molecular Formula Na2HPO4
Formula Weight 141.96
Synonym (Disodium hydrogen phosphate)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19473901

 
mp biomedicals
19473901
1kg Each for $156.23
Description & Specifications

Specifications

Purity Grade Cell Culture Reagent Grade
Formula Weight 141.96
Quantity 1kg
Synonym (Disodium hydrogen phosphate)
Chemical Name or Material Sodium Phosphate Dibasic
Name Note Anhydrous
Molecular Formula Na2HPO4
CAS 7558-79-4
Recommended Storage Room Temperature
Safety and Handling

Room Temperature